Please login to your online account to display your discounted pricing

GW 627368, Tocris™

Selective EP4 receptor competitive antagonist

$310.33 - $1,282.40

Chemical Identifiers

Chemical Name or Material GW 627368
Assay Percent Range >98%
Molecular Formula C30H28N2O6S
Formula Weight 544.62
Synonym 4-(4-, 9-Diethoxy-1, 3-dihydro-1-oxo-2H-benz[f]isoindol-2-yl)-N-(phenylsulfonyl)benzeneacetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

466810

 
r & d systems
RDS466810
10mg Each for $310.33

466850

 
r & d systems
RDS466850
50mg Each for $1,282.40
Description & Specifications

Specifications

Formula Weight 544.62
Quantity 10mg
Chemical Name or Material GW 627368
Assay Percent Range >98%
CAS 439288-66-1
Recommended Storage Store at -20°C
Synonym 4-(4-, 9-Diethoxy-1, 3-dihydro-1-oxo-2H-benz[f]isoindol-2-yl)-N-(phenylsulfonyl)benzeneacetamide
Molecular Formula C30H28N2O6S
Safety and Handling

Store at -20°C