Please login to your online account to display your discounted pricing

GW 542573X, Tocris™

Selective activator of KCa 2.1 channels

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material GW 542573X
Assay Percent Range >99%
Molecular Formula C19H28N2O5
Formula Weight 364.44
Synonym 4-[[[[(2-Methoxyphenyl)amino]carbonyl]oxy]methyl]-piperidinecarboxylic acid-1, 1-dimethylethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

431110

 
r & d systems
RDS431110
10mg Each for $239.63

431150

 
r & d systems
RDS431150
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 364.44
Quantity 10mg
Chemical Name or Material GW 542573X
Assay Percent Range >99%
Molecular Formula C19H28N2O5
CAS 660846-41-3
Synonym 4-[[[[(2-Methoxyphenyl)amino]carbonyl]oxy]methyl]-piperidinecarboxylic acid-1, 1-dimethylethyl ester
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C