Please login to your online account to display your discounted pricing

GW 3965 hydrochloride, Tocris

Orally active liver X receptor (LXR) agonist

$146.74 - $316.20

Chemical Identifiers

Chemical Name or Material GW 3965 hydrochloride
Assay Percent Range >98%
Molecular Formula C33H31NO3ClF3·HCl
Formula Weight 618.51
Synonym 3-[3-[[[2-Chloro-3-(trifluoromethyl)phenyl]methyl](2, 2-diphenylethyl)amino]propoxy]benzeneacetic acid hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

24741

 
r & d systems
24741
1mg Each for $146.74

247410

 
r & d systems
247410
10mg Each for $316.20
Description & Specifications

Specifications

Formula Weight 618.51
Quantity 1mg
Chemical Name or Material GW 3965 hydrochloride
Assay Percent Range >98%
Molecular Formula C33H31NO3ClF3·HCl
CAS 405911-17-3
Synonym 3-[3-[[[2-Chloro-3-(trifluoromethyl)phenyl]methyl](2, 2-diphenylethyl)amino]propoxy]benzeneacetic acid hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature