Please login to your online account to display your discounted pricing

GW 0742, Tocris

Highly selective, potent PPARδ agonist

$246.52 - $1,023.37

Chemical Identifiers

Chemical Name or Material GW 0742
Assay Percent Range >98%
Molecular Formula C21H17F4NO3S2
Formula Weight 471.49
Synonym [4-[[[2-[3-Fluoro-4-(trifluoromethyl)phenyl]-4-methyl-5-thiazolyl]methyl]thio]-2-methylphenoxy]acetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

222910

 
r & d systems
RDS222910
10mg Each for $246.52

222950

 
r & d systems
RDS222950
50mg Each for $1,023.37
Description & Specifications

Specifications

Formula Weight 471.49
Quantity 10mg
Chemical Name or Material GW 0742
CAS 317318-84-6
Synonym [4-[[[2-[3-Fluoro-4-(trifluoromethyl)phenyl]-4-methyl-5-thiazolyl]methyl]thio]-2-methylphenoxy]acetic acid
Molecular Formula C21H17F4NO3S2
Assay Percent Range >98%
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C