GVS Maine Polypropylene Syringe Filters 30mm
For aqueous and organic solvent-based samples
$218.15 - $2,162.28
Specifications
Applications Aqueous and organic solvent-based samples
Diameter (Metric) 30mm
Type Syringe Filter
Hold up Volume (After Air Purge) <60μL
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Material Pore Size Quantity Price Quantity  
Catalog Number Mfr. No. Material Pore Size Quantity Price Quantity  
DDP02T30NM GVS Maine
MM1224173
PolySep Polypropylene (PP) 0.22μm 200/Pk. Pack of 200 Each for $764.30
DDP02T3050 GVS Maine
MM1224172
PolySep Polypropylene (PP) 0.22μm 50/pk. Pack of 50 Each for $218.15
DDP04T301K GVS Maine
MM1229458
PolySep Polypropylene (PP) 0.45μm 1000/Pk. Pack of 1000 Each for $2,162.28
DDP04T3050 GVS Maine
MM1224310
PolySep Polypropylene (PP) 0.45μm 50/pk. Pack of 50 Each for $218.15
DDP04T30NB GVS Maine
MM1224312
PolySep Polypropylene (PP) 0.45μm 500/Pk. Pack of 500 Each for $1,708.15
DDP04T30NM GVS Maine
MM1224311
PolySep Polypropylene (PP) 0.45μm 200/Pk. Pack of 200 Each for $764.30
DGP02T3050 GVS Maine
MM1224175
PolySep Polypropylene with Binderless Glass Prefilter 0.22μm 50/pk. Pack of 50 Each for $233.00
DGP04T3050 GVS Maine
MM1224313
PolySep Polypropylene with Binderless Glass Prefilter 0.45μm 50/pk. Pack of 50 Each for $233.00
DGP04T30NM GVS Maine
MM1224314
PolySep Polypropylene with Binderless Glass Prefilter 0.45μm 200/Pk. Pack of 200 Each for $825.13
Specifications
Description
  • Recommended for ion chromatography, HPLC sample preparation, and analysis requiring detection levels below 230nm.
    • Polypropylene membrane filter offers broad chemical compatibility
    • For aqueous and organic solvent-based samples
    • With or without glass prefilters
    • For 17mm syringe filters, see cat. no. DGP02T1750
    Manufactured by GVS Maine.
Description & Specifications
Specifications
Aqueous and organic solvent-based samples
30mm
Syringe Filter
<60μL