missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GVS Maine Polypropylene Syringe Filters 30mm

For aqueous and organic solvent-based samples

Manufacturer:  GVS Maine MM1224311

 View more versions of this product

Catalog No. DDP04T30NM


 

Specifications Description & Specifications

Specifications

Syringe Filter
PolySep Polypropylene (PP)
Aqueous and organic solvent-based samples
0.45μm
male Luer slip
30mm
130psi
<60μL
female Luer-Lok and Luer slip
200/Pk.