Please login to your online account to display your discounted pricing
GVS Maine Polypropylene Syringe Filters 30mm
For aqueous and organic solvent-based samples
Manufacturer: gvs maine  MM1224312
 View more versions of this product
Catalog No. DDP04T30NB
  • / Pack of 500 Each
Description & Specifications
Specifications
Quantity 500/Pk.
Pore Size 0.45μm
Diameter (Metric) 30mm
Material PolySep Polypropylene (PP)
Applications Aqueous and organic solvent-based samples
Hold up Volume (After Air Purge) <60μL
Type Syringe Filter