GVS Maine Polypropylene Syringe Filters 17mm
For aqueous and organic solvent-based samples
$145.43 - $1,147.85
Specifications
Applications Aqueous and organic solvent-based samples
Diameter (Metric) 17mm
Type Syringe Filter
Inlet Style female Luer-Lok and Luer slip
Pressure Rating 130psi
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Material Pore Size Quantity Price Quantity  
Catalog Number Mfr. No. Material Pore Size Quantity Price Quantity  
DDP02T1750 GVS Maine
MM1224808
PolySep Polypropylene (PP) 0.22μm Pk. of 50 Pack of 50 Each for $145.43
DDP02T17NM GVS Maine
MM1224809
PolySep Polypropylene (PP) 0.22μm Pk. of 200 Pack of 200 Each for $492.72
DDP02T17NB GVS Maine
MM1224810
PolySep Polypropylene (PP) 0.22μm Pk. of 500 Pack of 500 Each for $1,147.85
DDP04T1750 GVS Maine
MM1224811
PolySep Polypropylene (PP) 0.45μm Pk. of 50 Pack of 50 Each for $154.94
DDP04T17NB GVS Maine
MM1224813
PolySep Polypropylene (PP) 0.45μm Pk. of 500 Pack of 500 Each for $1,077.43
DDP04T17NM GVS Maine
MM1224812
PolySep Polypropylene (PP) 0.45μm Pk. of 200 Pack of 200 Each for $492.72
DGP02T1750 GVS Maine
MM1224814
PolySep Polypropylene with Binderless Glass Prefilter 0.22μm Pk. of 50 Pack of 50 Each for $157.30
DGP04T1750 GVS Maine
MM1224817
PolySep Polypropylene with Binderless Glass Prefilter 0.45μm Pk. of 50 Pack of 50 Each for $167.58
DGP04T17NM GVS Maine
MM1224818
PolySep Polypropylene with Binderless Glass Prefilter 0.45μm Pk. of 200 Pack of 200 Each for $528.32
Specifications
Description
  • Recommended for ion chromatography, HPLC sample preparation, and analysis requiring detection levels below 230nm.
    • Polypropylene membrane filter offers broad chemical compatibility
    • For aqueous and organic solvent-based samples
    • With or without glass prefilters
    • See cat. no. DGP02T3050 for 30mm filters
    Manufactured by GVS Maine.
Description & Specifications
Specifications
Aqueous and organic solvent-based samples
Syringe Filter
130psi
17mm
female Luer-Lok and Luer slip