Please login to your online account to display your discounted pricing

GVS Maine Cameo Celulose Acetate Syringe Filters: 25mm

Ideal for filtration of tissue culture media, biological fluids, protein, enzyme, hybridization buffer and other aqueous solutions

Manufacturer: gvs maine  MM1213678

 View all options for this product

Catalog No. DDA02025SM


Description & Specifications


Type Syringe Filter
Diameter (Metric) 25mm
Material AcetatePlus Cellulose Acetate
Pressure Rating 75psi
Hold up Volume (After Air Purge) <60μL
Filter Type Cameo 25AS
Inlet Style Female Luer Lok
Outlet Style Male Luer Slip
Temperature (English) 131°F
Temperature (Metric) 55°C
Quantity Pk. of 200
Pore Size 0.22μm
Sterile Yes
Tip Material Modified Acrylic
Pressure Rating (Metric) 5.2kg/cm sq.

  • Low hold-up volume, low protein binding
  • Choose from sterile and nonsterile models
  • Available in two sizes with cellulose acetate membrane

  • Filters for samples <100mL
  • Modified acrylic housing supports membrane bidirectionally for injection or aspiration
  • 100% integrity tested
  • For easy identification, pore size and filter labeled on each unit
  • Female locking Luer inlet; male slip Luer outlet
  • 3mm filters also available; see cat. no. DDA0200300