Please login to your online account to display your discounted pricing

GVS Maine Cameo Celulose Acetate Syringe Filters: 25mm

Ideal for filtration of tissue culture media, biological fluids, protein, enzyme, hybridization buffer and other aqueous solutions

$252.79 - $252.79

Specifications

Diameter (Metric) 25mm
Filter Type Cameo 25AS
Hold up Volume (After Air Purge) <60μL
Inlet Style Female Luer Lok
Material AcetatePlus Cellulose Acetate
View More Specs
Products
Catalog Number Mfr. No. Pore Size Price Quantity    

DDA08025S0

 
gvs maine
MM1213758
0.8μm Pack of 50 Each for $252.79
Description & Specifications

Specifications

Diameter (Metric) 25mm
Filter Type Cameo 25AS
Hold up Volume (After Air Purge) <60μL
Inlet Style Female Luer Lok
Material AcetatePlus Cellulose Acetate
Outlet Style Male Luer Slip
Pressure Rating 75psi
Pressure Rating (Metric) 5.2kg/cm sq.
Sterile Yes
Temperature (English) 131°F
Temperature (Metric) 55°C
Tip Material Modified Acrylic
Type Syringe Filter

  • Low hold-up volume, low protein binding
  • Choose from sterile and nonsterile models
  • Available in two sizes with cellulose acetate membrane

  • Filters for samples <100mL
  • Modified acrylic housing supports membrane bidirectionally for injection or aspiration
  • 100% integrity tested
  • For easy identification, pore size and filter labeled on each unit
  • Female locking Luer inlet; male slip Luer outlet
  • 3mm filters also available; see cat. no. DDA0200300