Please login to your online account to display your discounted pricing

GVS Maine Cameo Celulose Acetate Syringe Filters: 25mm


Ideal for filtration of tissue culture media, biological fluids, protein, enzyme, hybridization buffer and other aqueous solutions

Manufacturer: gvs maine  MM1213758

 View more versions of this product

Catalog No. DDA08025S0

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Hold up Volume (After Air Purge) <60μL
Tip Material Modified Acrylic
Filter Type Cameo 25AS
Temperature (Metric) 55°C
Quantity Pk. of 50
Sterile Yes
Pressure Rating (Metric) 5.2kg/cm sq.
Diameter (Metric) 25mm
Material AcetatePlus Cellulose Acetate
Pressure Rating 75psi
Inlet Style Female Luer Lok
Outlet Style Male Luer Slip
Temperature (English) 131°F
Pore Size 0.8μm
Type Syringe Filter