GVS Maine Glass/Nylon Syringe Filters: 30mm Diameter
$233.23 - $1,844.94
Specifications
Diameter (Metric) 30mm
Type Syringe Filter
Filtration Area (Metric) 4.8cm sq.
Material Nylon w/Binderless Glass Prefilter
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Pore Size Quantity Price Quantity  
Catalog Number Mfr. No. Pore Size Quantity Price Quantity  
DGR02T30NB GVS Maine
MM1224129
0.22μm Pk. of 500 Pack of 500 Each for $1,844.94
DGR02T30NM GVS Maine
MM1224128
0.22μm Pk. of 200 Pack of 200 Each for $825.92
DGR02T3050 GVS Maine
MM1224127
0.22μm Pk. of 50 Pack of 50 Each for $233.23
DGR04T3050 GVS Maine
MM1224135
0.45μm Pk. of 50 Pack of 50 Each for $233.23
DGR04T30NB GVS Maine
MM1224137
0.45μm Pk. of 500 Pack of 500 Each for $1,844.94
DGR04T30NM GVS Maine
MM1224136
0.45μm Pk. of 200 Pack of 200 Each for $825.92
Specifications
Description
Nylon provides high throughput and low extractable levels without wetting agents
  • Designed for preparation of viscous aqueous and solvent samples with high particulate loads
  • Nylon membrane filter contains no wetting agents and is prerinsed in methanol, yielding extremely low extractable levels
  • Compatible with most organic solvents, including acetone, methylene chloride and acetonitrile
Order Info
Available in two filter sizes: Cameo 17 (see cat. no. DGR02T1750) with a 17mm syringe filter for sample volume <18mL and Cameo 30 with a 30mm filter for sample volume <180mL.
Manufactured by GVS Maine.
Description & Specifications
Specifications
30mm
Syringe Filter
4.8cm sq.
Nylon w/Binderless Glass Prefilter