GVS Maine Glass/Nylon Syringe Filters: 17mm Diameter
$162.02 - $1,198.36
Specifications
Diameter (Metric) 17mm
Type Syringe Filter
Material Nylon w/Binderless Glass Prefilter
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Pore Size Quantity Price Quantity  
Catalog Number Mfr. No. Pore Size Quantity Price Quantity  
DGR02T1750 GVS Maine
MM1224766
0.22μm Pk. of 50 Pack of 50 Each for $162.02
DGR02T17NM GVS Maine
MM1224767
0.22μm Pk. of 200 Pack of 200 Each for $544.15
DGR04T1750 GVS Maine
MM1224773
0.45μm Pk. of 50 Pack of 50 Each for $162.02
DGR04T17NB GVS Maine
MM1224775
0.45μm Pk. of 500 Pack of 500 Each for $1,198.36
DGR04T17NM GVS Maine
MM1224774
0.45μm Pk. of 200 Pack of 200 Each for $544.15
Specifications
Description
Nylon provides high throughput and low extractable levels without wetting agents
  • Designed for preparation of viscous aqueous and solvent samples with high particulate loads
  • Nylon membrane filter contains no wetting agents and is prerinsed in methanol, yielding extremely low extractable levels
  • Compatible with most organic solvents, including acetone, methylene chloride and acetonitrile
Order Info
Available in two filter sizes: Cameo 17 with a 17mm syringe filter for sample volume <18mL and Cameo 30 (see cat. no. DGR02T3050) with a 30mm filter for sample volume <180mL.
Manufactured by GVS Maine.
Description & Specifications
Specifications
17mm
Syringe Filter
Nylon w/Binderless Glass Prefilter