Please login to your online account to display your discounted pricing

GVS Maine Sterile Cameo Cellulose Acetate Syringe Filters with Glass Prefilters

Ideal for preparing tissue culture media or clarifying biological, protein, enzyme, probe and hybridization buffer solutions

$231.87 - $875.81

Specifications

Applications Tissue culture media preparation. Sterile filtration/clarification of: biological fluids; protein, enzyme, probe solutions; hybridization buffers; other aqueous samples.
Diameter (Metric) 25mm
Filter Type Cameo 25GAS
Hold up Volume (After Air Purge) <60μL
Inlet Style Female Luer Lok
View More Specs
Products
Catalog Number Mfr. No. Pore Size Quantity Price Quantity    

DGA04025S0

 
gvs maine
MM1214358
0.45μm 50/Pk. Pack of 50 Each for $231.87

DGA04025SM

 
gvs maine
MM1214360
0.45μm 200/pk. Pack of 200 Each for $875.81

DGA08025SM

 
gvs maine
MM1213122
0.8μm 200/Pk. Pack of 200 Each for $875.81

DGA08025S0

 
gvs maine
MM1212630
0.8μm 50/Pk. Pack of 50 Each for $231.87
Description & Specifications

Specifications

Applications Tissue culture media preparation. Sterile filtration/clarification of: biological fluids; protein, enzyme, probe solutions; hybridization buffers; other aqueous samples.
Diameter (Metric) 25mm
Filter Type Cameo 25GAS
Hold up Volume (After Air Purge) <60μL
Inlet Style Female Luer Lok
Material Cellulose acetate
Outlet Style Male Luer Slip
Pressure Rating 75psi
Pressure Rating (Metric) 5.2kg/cm sq.
Sterile Yes
Temperature (English) 131°F
Temperature (Metric) 55°C
Tip Material Acrylic
Type Syringe Filter

  • 25mm syringe filters with borosilicate glass prefilters and cellulose acetate membranes
  • High throughput, low hold-up volume, negligible protein binding
  • Maximum sample recovery
  • 100% integrity tested acrylic housing
  • Pore size and membrane printed on each filter for easy identification
  • Female locking Luer inlet; male slip Luer outlet
  • Gamma radiation sterilized
  • Packaged in individual blister packs