Please login to your online account to display your discounted pricing

GVS Maine Sterile Cameo Cellulose Acetate Syringe Filters with Glass Prefilters


Ideal for preparing tissue culture media or clarifying biological, protein, enzyme, probe and hybridization buffer solutions

Manufacturer: gvs maine  MM1213122

 View more versions of this product

Catalog No. DGA08025SM

  • / Pack of 200 Each
 

Description & Specifications

Specifications

Tip Material Acrylic
Pressure Rating (Metric) 5.2kg/cm sq.
Material Cellulose acetate
Outlet Style Male Luer Slip
Quantity 200/Pk.
Temperature (Metric) 55°C
Hold up Volume (After Air Purge) <60μL
Temperature (English) 131°F
Diameter (Metric) 25mm
Sterile Yes
Inlet Style Female Luer Lok
Filter Type Cameo 25GAS
Pore Size 0.8μm
Pressure Rating 75psi
Applications Tissue culture media preparation. Sterile filtration/clarification of: biological fluids; protein, enzyme, probe solutions; hybridization buffers; other aqueous samples.
Type Syringe Filter