Please login to your online account to display your discounted pricing

GVS Maine Sterile Cameo Cellulose Acetate Syringe Filters with Glass Prefilters


Ideal for preparing tissue culture media or clarifying biological, protein, enzyme, probe and hybridization buffer solutions

Manufacturer: gvs maine  MM1212630

 View more versions of this product

Catalog No. DGA08025S0

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Hold up Volume (After Air Purge) <60μL
Filter Type Cameo 25GAS
Inlet Style Female Luer Lok
Outlet Style Male Luer Slip
Temperature (English) 131°F
Temperature (Metric) 55°C
Quantity 50/Pk.
Pore Size 0.8μm
Sterile Yes
Tip Material Acrylic
Pressure Rating (Metric) 5.2kg/cm sq.
Type Syringe Filter
Diameter (Metric) 25mm
Material Cellulose acetate
Pressure Rating 75psi
Applications Tissue culture media preparation. Sterile filtration/clarification of: biological fluids; protein, enzyme, probe solutions; hybridization buffers; other aqueous samples.