Please login to your online account to display your discounted pricing

GVS Maine Sterile Cameo Cellulose Acetate Syringe Filters with Glass Prefilters

Ideal for preparing tissue culture media or clarifying biological, protein, enzyme, probe and hybridization buffer solutions

Manufacturer: gvs maine  MM1214359

 View all options for this product

Catalog No. DGA04025SB


Description & Specifications


Type Syringe Filter
Diameter (Metric) 25mm
Material Cellulose acetate
Pressure Rating 75psi
Applications Tissue culture media preparation. Sterile filtration/clarification of: biological fluids; protein, enzyme, probe solutions; hybridization buffers; other aqueous samples.
Hold up Volume (After Air Purge) <60μL
Filter Type Cameo 25GAS
Inlet Style Female Luer Lok
Outlet Style Male Luer Slip
Temperature (English) 131°F
Temperature (Metric) 55°C
Quantity 500/Pk.
Pore Size 0.45μm
Sterile Yes
Tip Material Acrylic
Pressure Rating (Metric) 5.2kg/cm sq.

  • 25mm syringe filters with borosilicate glass prefilters and cellulose acetate membranes
  • High throughput, low hold-up volume, negligible protein binding
  • Maximum sample recovery
  • 100% integrity tested acrylic housing
  • Pore size and membrane printed on each filter for easy identification
  • Female locking Luer inlet; male slip Luer outlet
  • Gamma radiation sterilized
  • Packaged in individual blister packs