GVS Polyvinylidene Fluoride Hybridization and Transfer Membranes
Designed for Western transfers, amino acid analyses, and protein sequencing procedures
Supplier:  GVS 1214558
 View more versions of this product
Catalog No. PV4HY910F0
Description
Description
  • Compatible with most commonly used stains
  • Won't degrade, distort or shrink in presence of high concentrations of methanol
  • Efficient protein transfer from 5 to 700kDa
  • Detection limit, 0.005μg/cm2
  • Packaged in a wide variety of precut circles and sheets to fit Petri dishes and transfer/hybridization apparatus
  • Also available in rolls for cutting custom-sized membranes
Manufactured by GVS Maine.
Specifications
Specifications
9 cm
For Western transfers, amino acid analysis, protein sequencing
Sheet
0.45 μm
10 cm
125 μg/cm2
PVDF-Plus Polyvinylidene Fluoride