GVS Maine Nylon Syringe Filters
Compatible with acetone, methylene chloride, acetonitrile and other organic solvents
$158.38 - $1,853.04
Specifications
Type Syringe Filter
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Housing Material Pore Size Quantity Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Housing Material Pore Size Quantity Diameter (Metric) Price Quantity  
DDR01T3050 GVS Maine
MM1224100
Polypropylene 0.1μm 50/Pk. 30mm Pack of 50 Each for $237.57
DDR01T30NB GVS Maine
MM1224103
Polypropylene 0.1μm 500/Pk. 30mm Pack of 500 Each for $1,746.02
DDR02T1750 GVS Maine
MM1224746
Polypropylene 0.22μm 50/Pk. 17mm Pack of 50 Each for $158.38
DDR02T17NB GVS Maine
MM1224748
Polypropylene 0.22μm 500/Pk. 17mm Pack of 500 Each for $1,101.30
DDR02T17NM GVS Maine
MM1224747
Polypropylene 0.22μm 200/Pk. 17mm Pack of 200 Each for $503.64
DDR02T3050 GVS Maine
MM1224104
Polypropylene 0.22μm 50/Pk. 30mm Pack of 50 Each for $254.77
DDR02T30NB GVS Maine
MM1224106
Polypropylene 0.22μm 500/Pk. 30mm Pack of 500 Each for $1,746.02
DDR02T30NM GVS Maine
MM1224105
Polypropylene 0.22μm 200/Pk. 30mm Pack of 200 Each for $781.25
DDR04T17NB GVS Maine
MM1224755
Polypropylene 0.45μm 500/Pk. 17mm Pack of 500 Each for $1,101.30
DDR04T1750 GVS Maine
MM1224753
Polypropylene 0.45μm 50/Pk. 17mm Pack of 50 Each for $158.38
DDR04T30NB GVS Maine
MM1224114
Polypropylene 0.45μm 500/Pk. 30mm Pack of 500 Each for $1,853.04
DDR04T17NM GVS Maine
MM1224754
Polypropylene 0.45μm 200/Pk. 17mm Pack of 200 Each for $503.64
DDR04T3050 GVS Maine
MM1224112
Polypropylene 0.45μm 50/Pk. 30mm Pack of 50 Each for $223.00
DDR12T3050 GVS Maine
MM1224119
Polypropylene 1.2μm 50/Pk. 30mm Pack of 50 Each for $237.57
DDR12T30NM GVS Maine
MM1224120
Polypropylene 1.2μm 200/Pk. 30mm Pack of 200 Each for $781.25
DDR5000300 GVS Maine
MM1213742
Virgin Nylon 66 5μm 100/Pk. 3.0mm Pack of 100 Each for $221.42
DDR50T3050 GVS Maine
MM1224124
Polypropylene 5μm 50/Pk. 30mm Pack of 50 Each for $237.57
DDR50T30NM GVS Maine
MM1224125
Polypropylene 5μm 200/Pk. 30mm Pack of 200 Each for $781.25
DDR02025SM GVS Maine
MM1213711
Modified Acrylic 0.22μm 200/Pk. 25mm Pack of 200 Each for $781.25
DDR50T30NB GVS Maine
MM1224126
Polypropylene 5μm 500/Pk. 30mm Pack of 500 Each for $1,746.02
Specifications
Description
  • Disposable Inherently hydrophilic Magna™ Nylon membranes contain no wetting agents
  • Yield exceptionally low extractable levels (<0.0015mg/cm2) Female Luer-Lok™ and slip Luer inlet; male slip Luer outlet
  • Autoclavable
  • Available in nonsterile or sterilized packaging
3mm dia. (3R) Filters
  • For direct injection of aqueous and organic samples of 1mL or less into sample loop systems
  • Virgin Nylon 66 housing resists heat, common aggressive solvents
17mm dia. (17R) Filters
  • Designed to filter aqueous and organic samples of 12mL or less directly injected into HPLC loop systems or other small sample preparation
  • Filtered samples ensure column protection, trouble-free operation, consistent test results
  • Polypropylene housing
25mm dia. (25RS) Filters
  • Sterile
  • For aqueous samples including biological fluids, buffer solutions, tissue culture media
  • Modified acrylic housing supports membrane bidirectionally for injection or aspiration
  • 100% integrity tested
  • Filter and pore size printed on housing
  • In individual blister packs
30mm dia. (30R) Filters
  • Designed for sterilization or clarification of aqueous and solvent samples up to 120mL in a wide array of analytical and research applications
  • Allow direct injection of sample into HPLC loop systems
  • Polypropylene housing
Manufactured by GE Water & Process Technologies.
Description & Specifications
Specifications
Syringe Filter