GVS Maine Nylon Syringe Filters
Compatible with acetone, methylene chloride, acetonitrile and other organic solvents
Manufacturer:  GVS Maine MM1224126
 View more versions of this product
Catalog No. DDR50T30NB
Specifications Description & Specifications
Specifications
Syringe Filter
MAGNA Nylon
Polypropylene
4.8cm sq.
male Luer slip
30mm
For sterilization or clarification of aqueous and solvent samples
5μm
female Luer-Lok and Luer slip
500/Pk.