Please login to your online account to display your discounted pricing
GVS Maine Nylon Syringe Filters
Compatible with acetone, methylene chloride, acetonitrile and other organic solvents
Manufacturer: gvs maine  MM1213711
 View more versions of this product
Catalog No. DDR02025SM
  • / Pack of 200 Each
Description & Specifications
Specifications
Housing Material Modified Acrylic
Pore Size 0.22μm
Diameter (Metric) 25mm
Quantity 200/Pk.
Type Syringe Filter
Material Nylon (NYL)
Applications For aqueous samples such as biological fluids, buffer solutions, tissue culture media