GVS Maine Nylon Syringe Filters
Compatible with acetone, methylene chloride, acetonitrile and other organic solvents
Manufacturer:  GVS Maine MM1224747
 View more versions of this product
Catalog No. DDR02T17NM
Specifications Description & Specifications
Specifications
Polypropylene
0.22μm
Nylon (NYL)
200/Pk.
To filter aqueous and organic samples of 12mL or less
male Luer slip
1.0cm sq.
17mm
female Luer-Lok and Luer slip
Syringe Filter