missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
GVS Nylon Syringe Filters
Compatible with acetone, methylene chloride, acetonitrile and other organic solvents
Supplier: GVS 1224120
Description
- Disposable Inherently hydrophilic Magna™ Nylon membranes contain no wetting agents
- Yield exceptionally low extractable levels (<0.0015mg/cm2) Female Luer-Lok™ and slip Luer inlet; male slip Luer outlet
- Autoclavable
- Available in nonsterile or sterilized packaging
3mm dia. (3R) Filters
- For direct injection of aqueous and organic samples of 1mL or less into sample loop systems
- Virgin Nylon 66 housing resists heat, common aggressive solvents
- Designed to filter aqueous and organic samples of 12mL or less directly injected into HPLC loop systems or other small sample preparation
- Filtered samples ensure column protection, trouble-free operation, consistent test results
- Polypropylene housing
- Sterile
- For aqueous samples including biological fluids, buffer solutions, tissue culture media
- Modified acrylic housing supports membrane bidirectionally for injection or aspiration
- 100% integrity tested
- Filter and pore size printed on housing
- In individual blister packs
- Designed for sterilization or clarification of aqueous and solvent samples up to 120mL in a wide array of analytical and research applications
- Allow direct injection of sample into HPLC loop systems
- Polypropylene housing
Manufactured by GE Water & Process Technologies.
Specifications
MAGNA Nylon | |
4.8 cm2 | |
Polypropylene | |
male Luer slip | |
Syringe Filter |
1.2 μm | |
For sterilization or clarification of aqueous and solvent samples | |
female Luer-Lok and Luer slip | |
200/Pk. | |
30 mm |