Please login to your online account to display your discounted pricing

GVS Maine Nylon Syringe Filters

Compatible with acetone, methylene chloride, acetonitrile and other organic solvents

Manufacturer: gvs maine  MM1213724

 View all options for this product

Catalog No. DDR04025S0


Description & Specifications


Type Syringe Filter
Diameter (Metric) 25mm
Material MAGNA Nylon
Pore Size 0.45μm
Quantity 50/Pk.

  • Disposable Inherently hydrophilic Magna™ Nylon membranes contain no wetting agents
  • Yield exceptionally low extractable levels (<0.0015mg/cm2) Female Luer-Lok™ and slip Luer inlet; male slip Luer outlet
  • Autoclavable
  • Available in nonsterile or sterilized packaging

3mm dia. (3R) Filters

  • For direct injection of aqueous and organic samples of 1mL or less into sample loop systems
  • Virgin Nylon 66 housing resists heat, common aggressive solvents
17mm dia. (17R) Filters
  • Designed to filter aqueous and organic samples of 12mL or less directly injected into HPLC loop systems or other small sample preparation
  • Filtered samples ensure column protection, trouble-free operation, consistent test results
  • Polypropylene housing
25mm dia. (25RS) Filters
  • Sterile
  • For aqueous samples including biological fluids, buffer solutions, tissue culture media
  • Modified acrylic housing supports membrane bidirectionally for injection or aspiration
  • 100% integrity tested
  • Filter and pore size printed on housing
  • In individual blister packs
30mm dia. (30R) Filters
  • Designed for sterilization or clarification of aqueous and solvent samples up to 120mL in a wide array of analytical and research applications
  • Allow direct injection of sample into HPLC loop systems
  • Polypropylene housing