GVS Nitrocellulose-Mixed Esters of Cellulose Membrane Filters
Uniform microporous structure has low extractables, low ash
Manufacturer:  GVS MM1215412
 View more versions of this product
Catalog No. E08BG04700
Description
Description
Order Info
In dispenser packs of 25 or 100. Nitrocellulose 47mm membrane filters in 0.22 and 0.45μm thickness are also available in presterilized packs, as well as autoclavable packs suitable for onsite presterilization. Order online or contact your Fisher Scientific™ Customer Service Representative.
Specifications
Specifications
0.8μm
47mm
15psi
17.0L/min./cm sq.
150μm
Grid
Mixed Cellulose Esters (MCE) (Nitrate and Acetate)
Black
Microbiological determinations; industrial hygiene procedures; clarifying aqueous solutions; determination of lead, chromium, cadmium, beryllium, arsenic, airborne asbestos and other particulates.
195.0mL/min./cm sq.
Hydrophilic
Documents