GVS Nitrocellulose Hybridization and Transfer Membranes: Sheets (45μm)
Require vacuum baking to fix DNA
Manufacturer:  GVS MM1215478
 View more versions of this product
Catalog No. EP4HY322F5
Description
Description
Nitrocellulose Unsupported Membrane (EP)
  • Highest sensitivity for protein blotting and immunoblotting, or other procedures requiring optimum resolution
  • Easily blocked
  • Wets out naturally and does not require methanol like PVDF membranes
Nitrocellulose Supported Membrane (WP)
  • High sensitivity but with added strength of a polyester matrix
  • Surfactant-free membranes have no additives to interfere with binding capacity
Specifications
Specifications
8.6 in.
8.6 in.
NitroBind
0.45μm
22cm
22cm
Sheet
Documents