missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GVS Nitrocellulose Hybridization and Transfer Membranes: Sheets (45μm)

Require vacuum baking to fix DNA

Manufacturer:  GVS MM1215477

 View more versions of this product

Catalog No. EP4HY320F5



Description

Description

Nitrocellulose Unsupported Membrane (EP)

  • Highest sensitivity for protein blotting and immunoblotting, or other procedures requiring optimum resolution
  • Easily blocked
  • Wets out naturally and does not require methanol like PVDF membranes
Nitrocellulose Supported Membrane (WP)
  • High sensitivity but with added strength of a polyester matrix
  • Surfactant-free membranes have no additives to interfere with binding capacity

Specifications

Specifications

7.8 in.
7.8 in.
For protein blotting and immunoblotting, or other procedures requiring optimum resolution
Sheet
20cm
20cm
NitroBind
0.45μm
Documents