Please login to your online account to display your discounted pricing

GVS Maine Nitrocellulose Hybridization and Transfer Membranes: Circles


Require vacuum baking to fix DNA

Manufacturer: gvs maine  WP4HY08250

 View more versions of this product

Catalog No. WP4HY08250

  • / Pack of 50 Each
 

Description & Specifications

Specifications

For Use With (Application) Transfer Membrane
Material NitroPure
Diameter (Metric) 82mm
Pore Size 0.45μm
Format Circle