GVS Neutral Nylon Membranes
Do not require prewetting or vacuum baking
$193.00 - $807.45
Specifications
Material Neutral Nylon
Pore Size 0.45 μm
View More Specs
Products 6
Catalog Number Mfr. No. Description Format Quantity Price Quantity  
Catalog Number Mfr. No. Description Format Quantity Price Quantity  
N00HY08250
View Documents
GVS
1213370
82mm dia. Circle 50/Pk.
Pack of 50 Each for $358.21
N00HY08550
View Documents
GVS
1213372
85mm dia. Circle 50/Pk.
Pack of 50 Each for $369.61
N00HY315F5
View Documents
GVS
1213379
15 x 15cm Sheet 5/Pk.
Pack of 5 Each for $193.00
N00HYB0010
View Documents
GVS
1213404
15cm x 3m Roll 1 Each
Each for $420.76
N00HYA0010
View Documents
GVS
1213403
20cm x 3m Roll 1 Each
Each for $551.55
N00HY00010
View Documents
GVS
1213364
30cm x 3m Roll 1 Each
Each for $807.45
Description
Description
  • Packaged in a wide variety of precut circles and sheets to fit Petri dishes and transfer/hybridization apparatus
  • Also available in rolls for cutting custom-sized membranes
Nylon, Neutral (N0)
  • Supported nylon membrane
  • Ideal for colony/plaque lifts and single-probe applications
  • Low background yields clear autoradiographs
†Also available in 0.22μm pore size. Contact your Fisher Scientific™ Customer Service Representative.
Specifications
Specifications
Neutral Nylon
0.45 μm