GVS Neutral Nylon Membranes
Do not require prewetting or vacuum baking
Supplier: GVS 1213404
Description
- Packaged in a wide variety of precut circles and sheets to fit Petri dishes and transfer/hybridization apparatus
- Also available in rolls for cutting custom-sized membranes
Nylon, Neutral (N0)
- Supported nylon membrane
- Ideal for colony/plaque lifts and single-probe applications
- Low background yields clear autoradiographs
†Also available in 0.22μm pore size. Contact your Fisher Scientific™ Customer Service Representative.
Specifications
15 cm | |
15cm x 3m | |
Roll | |
0.45 μm |
3 m | |
1 Each | |
Neutral Nylon |