GVS Magna™ Nylon Membrane Filters
Plain white nylon membranes offer low extractables, high uniformity
Manufacturer:  GVS MM1213760
 View more versions of this product
Catalog No. R01SP01300
Description
Description
Plain white nylon membranes offer low extractables, high uniformity
  • Won't crack, tear, or distort in handling or creasing
  • Resist heat, sterilization, and most organic solvents
  • Hydrophilic, no wetting agents; extractable level <0.0015mg/cm
  • Heatable, autoclavable up to 180°C (356°F)
  • Broad chemical compatibility
Not compatible with strong acids or bases
  • Permit use for aqueous filtration without wetting and organic solvent filtrations such as HPLC
  • For microbiological work, use 47mm/0.45μm size with grid
†Grid-style membranes.
Specifications
Specifications
0.1μm
13mm
70psi
0.6L/min./cm sq.
Hydrophilic
Nylon (NYL)
Yes
Permit use for aqueous filtration without wetting and organic solvent filtrations such as HPLC
5.2mL/min./cm sq.
Plain
Documents