GVS Magna™ Nylon Membrane Filters
Plain white nylon membranes offer low extractables, high uniformity
$180.06 - $748.30
Specifications
Autoclavable | Yes |
---|---|
Material | Nylon (NYL) |
For Use With (Application) | Permit use for aqueous filtration without wetting and organic solvent filtrations such as HPLC |
Catalog Number | Mfr. No. | Diameter (Metric) Outer | Pore Size | Membrane Type | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Outer | Pore Size | Membrane Type | Price | Quantity | |||
R01SP09025
|
GVS
MM1213763 |
90mm | 0.1μm | Plain |
Pack of 25 Each for $341.82
|
|
R01SP04700
|
GVS
MM1213762 |
47mm | 0.1μm | Plain |
Pack of 100 Each for $366.85
|
|
R01SP02500
|
GVS
MM1213761 |
25mm | 0.1μm | Plain |
Pack of 100 Each for $289.06
|
|
R01SP01300
|
GVS
MM1213760 |
13mm | 0.1μm | Plain |
Pack of 100 Each for $225.12
|
|
R02SP04700
|
GVS
MM1213769 |
47mm | 0.2μm | Plain |
Pack of 100 Each for $318.36
|
|
R02SP01300
|
GVS
MM1213766 |
13mm | 0.2μm | Plain |
Pack of 100 Each for $197.33
|
|
R02SP02500
|
GVS
MM1213768 |
25mm | 0.2μm | Plain |
Pack of 100 Each for $234.70
|
|
R02SP09025
|
GVS
MM1213770 |
90mm | 0.2μm | Plain |
Pack of 25 Each for $188.97
|
|
R02SP29325
|
GVS
MM1213772 |
293mm | 0.2μm | Plain |
Pack of 25 Each for $748.30
|
|
R04SG04700
|
GVS
MM1213825 |
47mm | 0.5μm | Grid |
Pack of 100 Each for $341.82
|
|
R04SP01300
|
GVS
MM1213774 |
13mm | 0.5μm | Plain |
Pack of 100 Each for $197.33
|
|
R04SP09025
|
GVS
MM1213778 |
90mm | 0.45μm | Plain |
Pack of 25 Each for $197.33
|
|
R04SP02500
|
GVS
MM1213775 |
25mm | 0.5μm | Plain |
Pack of 100 Each for $234.70
|
|
R04SP04700
|
GVS
MM1213776 |
47mm | 0.5μm | Plain |
Pack of 100 Each for $327.94
|
|
R04SP14225
|
GVS
MM1213779 |
142mm | 0.5μm | Plain |
Pack of 25 Each for $391.88
|
|
R06SP04700
|
GVS
MM1213783 |
47mm | 0.7μm | Plain |
Pack of 100 Each for $366.85
|
|
R08SP02500
|
GVS
MM1213789 |
25mm | 0.8μm | Plain |
Pack of 100 Each for $265.00
|
|
R08SP04700
|
GVS
MM1213790 |
47mm | 0.8μm | Plain |
Pack of 100 Each for $366.85
|
|
R08SP09025
|
GVS
MM1213791 |
90mm | 0.8μm | Plain |
Pack of 25 Each for $289.06
|
|
R08SP14225
|
GVS
MM1213792 |
142mm | 0.8μm | Plain |
Pack of 25 Each for $391.88
|
|
R12SP01300
|
GVS
MM1213794 |
13mm | 1.2μm | Plain |
Pack of 100 Each for $197.33
|
|
R12SP02500
|
GVS
MM1213796 |
25mm | 1.2μm | Plain |
Pack of 100 Each for $245.15
|
|
R12SP09025
|
GVS
MM1213798 |
90mm | 1.2μm | Plain |
Pack of 25 Each for $305.70
|
|
R12SP14225
|
GVS
MM1213799 |
142mm | 1.2μm | Plain |
Pack of 25 Each for $325.15
|
|
R12SP04700
|
GVS
MM1213797 |
47mm | 1.2μm | Plain |
Pack of 100 Each for $356.18
|
|
R22SP01300
|
GVS
MM1213801 |
13mm | 20μm | Plain |
Pack of 100 Each for $180.06
|
|
R22SP09025
|
GVS
MM1213807 |
90mm | 20μm | Plain |
Pack of 25 Each for $278.97
|
|
R22SP14225
|
GVS
MM1213808 |
142mm | 20μm | Plain |
Pack of 25 Each for $355.12
|
|
R50SP02500
|
GVS
MM1213811 |
25mm | 5μm | Plain |
Pack of 100 Each for $241.79
|
|
R50SP04700
|
GVS
MM1213812 |
47mm | 5μm | Plain |
Pack of 100 Each for $366.85
|
|
R50SP01300
|
GVS
MM1213810 |
13mm | 5μm | Plain |
Pack of 100 Each for $197.33
|
|
R50SP14225
|
GVS
MM1213815 |
142mm | 5μm | Plain |
Pack of 25 Each for $358.55
|
|
R99SP01300
|
GVS
MM1213817 |
13mm | 10μm | Plain |
Pack of 100 Each for $221.48
|
|
R50SP09025
|
GVS
MM1213813 |
90mm | 5μm | Plain |
Pack of 25 Each for $305.70
|
|
R02SP14225
|
GVS
MM1213771 |
142mm | 0.2μm | Plain |
Pack of 25 Each for $441.15
|
|
Description
Plain white nylon membranes offer low extractables, high uniformity
- Won't crack, tear, or distort in handling or creasing
- Resist heat, sterilization, and most organic solvents
- Hydrophilic, no wetting agents; extractable level <0.0015mg/cm
- Heatable, autoclavable up to 180°C (356°F)
- Broad chemical compatibility
Not compatible with strong acids or bases
- Permit use for aqueous filtration without wetting and organic solvent filtrations such as HPLC
- For microbiological work, use 47mm/0.45μm size with grid
†Grid-style membranes.
Specifications
Yes | |
Nylon (NYL) | |
Permit use for aqueous filtration without wetting and organic solvent filtrations such as HPLC |