GVS Magna™ Nylon Membrane Filters
Plain white nylon membranes offer low extractables, high uniformity
Manufacturer:  GVS MM1213796
 View more versions of this product
Catalog No. R12SP02500
Description
Description
Plain white nylon membranes offer low extractables, high uniformity
†Grid-style membranes.
Specifications
Specifications
1.2μm
25mm
Yes
Permit use for aqueous filtration without wetting and organic solvent filtrations such as HPLC
190.0mL/min./cm sq.
Plain
Nylon (NYL)
100/pk.
10psi
18.0L/min./cm sq.
Hydrophilic
Documents