GVS Glass Prefilters
With or without binders
Supplier: GVS 1215588
Description
With or without binders
G40 Binder-free Filters
- High solvent resistance
- Higher dirt-handling capacity than glass filters with binders
- Made of borosilicate glass fibers woven into a porous matrix
- Ideal for gravimetric analyses and particulate testing
- Borosilicate glass fiber matrix bonded by acrylic resin
- G25 filters are extra thick for increased throughput
- High wet strength for long aqueous filtrations and reduced media migration
- Surface capture of particles
- Average retention efficiency
- Higher dirt-handling capacity
Specifications
Autoclavable | |
1μm | |
General purpose, routine filtrations, solvent filtrations | |
950°F | |
0.014 in. | |
1.4 in. |
Borosilicate Glass Fiber Woven into a Porous Matrix | |
Sterile | |
G40 | |
510°C | |
0.38 mm | |
3.7 cm |