Please login to your online account to display your discounted pricing

GVS Maine FAST™ Slide Incubation Chambers


Desgned with gasket to form tight and leak-proof seal

Manufacturer: gvs maine  MM10486046

 View more versions of this product

Catalog No. 10-486-046

  • / Pack of 10 Each
 

Description & Specifications

Specifications

For Use With (Equipment) FAST™ Slides
Format Single well
Quantity 10/Pk.
Type Incubation chamber

  • Used in conjunction with FAST Frame or Chip Clip™ Slide Holder
  • Conducts binding reactions on protein microarrays
  • Designed to fit all FAST Slide formats (1-, 2-, 8- and 16-pad)
  • With sensitivity and dynamic range
  • Provides long-term stability of printed proteins
  • Compatible with all detection methodologies
  • Compatible with commercially available arraying robots