Please login to your online account to display your discounted pricing

GVS Maine Cameo™ Syringe Filter Glass Fiber/Nylon


Manufacturer: gvs maine  MM1226916

 View more versions of this product

Catalog No. DGR04T301K

  • / Pack of 1000 Each
 

Description & Specifications

Specifications

Diameter (Metric) Inner 30mm
Sterile No
Description Syringe filter
Pore Size 0.45μm
Material Glass Fiber/Nylon (Membrane)
Autoclavable Yes
Color Transparent