GVS Maine Cameo PTFE Syringe Filters with Glass Prefilters: 30mm Diameter
For use with aggressive acids, bases and for degassing solvents
Manufacturer:  GVS Maine MM1224165
 View more versions of this product
Catalog No. DGF04T30NM
Specifications Description & Specifications
Specifications
0.45μm
TefSep PTFE with Borosilicate Glass Prefilter
Syringe Filter
4.8cm sq.
30mm