Please login to your online account to display your discounted pricing
GVS Maine Cameo PTFE Syringe Filters with Glass Prefilters: 30mm Diameter
For use with aggressive acids, bases and for degassing solvents
$298.30 - $879.94
Specifications
Diameter (Metric) 30mm
Type Syringe Filter
Material TefSep PTFE with Borosilicate Glass Prefilter
View More Specs
Products
Catalog Number Mfr. No. Filtration Area (Metric) Pore Size Price Quantity    
DGF04T3050
gvs maine
MM1224164
4.8cm sq. 0.45μm Pack of 50 Each for $298.30
DGF04T30NM
gvs maine
MM1224165
4.8cm sq. 0.45μm Pack of 200 Each for $879.94
Description & Specifications
Specifications
Diameter (Metric) 30mm
Type Syringe Filter
Material TefSep PTFE with Borosilicate Glass Prefilter
  • Naturally hydrophobic PTFE membrane filter will not release extractables into sample
  • For gas chromatography
  • Sterilizes or clarifies aggressive solvents, acids and bases
  • Use with vent filters to sterilize air, gases or aerosols