missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
GVS Cameo PTFE Syringe Filters with Glass Prefilters: 17mm Diameter
For use with aggressive acids, bases and for degassing solvents
Supplier: GVS 1224794
Description
- Naturally hydrophobic PTFE membrane filter will not release extractables into sample
- For HPLC solvent and gas chromatography
Specifications
TefSep with Binderless Glass Prefilter | |
1.0 cm2 | |
Pk. of 50 | |
17 mm |
0.22 μm | |
PTFE | |
Syringe Filter |