GVS Maine Cameo PTFE Syringe Filters with Glass Prefilters: 17mm Diameter
For use with aggressive acids, bases and for degassing solvents
$162.32 - $1,200.62
Specifications
Diameter (Metric) 17mm
Type Syringe Filter
Material TefSep with Binderless Glass Prefilter
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Pore Size Quantity Price Quantity  
Catalog Number Mfr. No. Pore Size Quantity Price Quantity  
DGF02T17NB GVS Maine
MM1224796
0.22μm Pk. of 500 Pack of 500 Each for $1,200.62
DGF02T1750 GVS Maine
MM1224794
0.22μm Pk. of 50 Pack of 50 Each for $171.47
DGF04T1750 GVS Maine
MM1224801
0.45μm Pk. of 50 Pack of 50 Each for $162.32
DGF04T17NB GVS Maine
MM1224803
0.45μm Pk. of 500 Pack of 500 Each for $1,200.62
Specifications
Description
  • Naturally hydrophobic PTFE membrane filter will not release extractables into sample
  • For HPLC solvent and gas chromatography
Description & Specifications
Specifications
17mm
Syringe Filter
TefSep with Binderless Glass Prefilter