Promega™ Gutathione Particles for MagneGST™ Protein Purification System DFS Item
$180.82 - $604.55
Specifications
For Use With (Application) Protein purification of Glutathione-S-transferase fusion Proteins from cleared or crude E. coli cell lysate, high-throughput purification of GST-tagged Proteins using auto mated platforms
Storage Requirements 4° or -70°C
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRV8612 Promega™
V8612
20mL Each for $604.55
PR-V8611 Promega™
V8611
4mL Each for $180.82
Specifications Description & Specifications
Specifications
Protein purification of Glutathione-S-transferase fusion Proteins from cleared or crude E. coli cell lysate, high-throughput purification of GST-tagged Proteins using auto mated platforms
4° or -70°C