missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gum guaiac, MP Biomedicals™

$1,464.87 - $1,464.87

Chemical Identifiers

CAS 9000-29-7
Physical Form Powder
Chemical Name or Material Gum guaiac
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15729191 Mp Biomedicals
0215729191
1kg Each for $1,464.87
Specifications Description & Specifications

Chemical Identifiers

9000-29-7
Powder
Gum guaiac

Specifications

9000-29-7
Light brown to greenish brown
Gum guaiac
Powder
1kg