Please login to your online account to display your discounted pricing

Guggulsterone, Tocris

Broad spectrum steroid receptor ligand; Antagonizes FXR and displays hypolipidemic activity

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material Guggulsterone
Assay Percent Range >99%
Molecular Formula C21H28O2
Formula Weight 312.45
Synonym Pregna-4, 17(20)-diene-3, 16-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

201310

 
r & d systems
RDS201310
10mg Each for $201.36

201350

 
r & d systems
RDS201350
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 312.45
Quantity 10mg
Chemical Name or Material Guggulsterone
Assay Percent Range >99%
CAS 95975-55-6
Synonym Pregna-4, 17(20)-diene-3, 16-dione
Molecular Formula C21H28O2
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature