Please login to your online account to display your discounted pricing

Gue 1654, Tocris™

Selective inhibitor of OXE-R Gβγ signaling

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material Gue 1654
Assay Percent Range >97%
Molecular Formula C23H17N3OS3
Formula Weight 447.6
Synonym N-[7-(Methylthio)benzo[1, 2-d:4, 3-d']bisthiazol-2-yl]-α-phenylbenzeneacetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

474210

 
r & d systems
RDS474210
10mg Each for $168.15

474250

 
r & d systems
RDS474250
50mg Each for $697.40
Description & Specifications

Specifications

Formula Weight 447.6
Quantity 10mg
Chemical Name or Material Gue 1654
Assay Percent Range >97%
Molecular Formula C23H17N3OS3
CAS 397290-30-1
Synonym N-[7-(Methylthio)benzo[1, 2-d:4, 3-d']bisthiazol-2-yl]-α-phenylbenzeneacetamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C