Please login to your online account to display your discounted pricing

Alfa Aesar™ Guanylurea phosphate, 98+%

$83.18 - $1,012.54

Chemical Identifiers

Chemical Name or Material Guanylurea phosphate
CAS 17675-60-4
Molecular Formula C6H18N12O3·H3PO4
Formula Weight 404.28
MDL Number MFCD00065097
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1853122

  SDS
alfa aesar™
A1853122
100g Each for $256.15

AAA1853114

  SDS
alfa aesar™
A1853114
25g Each for $83.18

AAA1853136

  SDS
alfa aesar™
A1853136
500g Each for $1,012.54
Description & Specifications

Specifications

Formula Weight 404.28
Quantity 100g
Molecular Formula C6H18N12O3·H3PO4
Chemical Name or Material Guanylurea phosphate
CAS 17675-60-4
MDL Number MFCD00065097
Assay ≥98%
TSCA TSCA
Melting Point 178° to 182°C