missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Guanosine 5'-monophosphate, disodium salt hydrate, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 226600050
Specifications
Guanosine 5′-monophosphate,disodium salt hydrate | |
25% max. (120°C, 4 hrs) | |
97% min. (lambda max. 252nm,pH 7) molar absorptivity 13700+/-4%,pH 7 | |
>300°C | |
5550-12-9 | |
C10H12N5Na2O8P·xH2O | |
10, 279 | |
5'-gmp disodium salt, 5'-guanylic acid disodium salt, 5'-guanylic acid, disodium salt, disodium 5'-gmp, disodium 5'-guanylate, disodium gmp, disodium guanylate, gmp disodium salt, guanosine 5'-monophosphate disodium salt, sodium guanylate | |
PVBRXXAAPNGWGE-LGVAUZIVSA-L | |
disodium;[(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate | |
21712 | |
Crystalline Powder |
Authentic | |
Glass bottle | |
White | |
5g | |
97% | |
MFCD00151238 | |
14, 4570 | |
Solubility in water: >50g/l water | |
C1=NC2=C(N1C3C(C(C(O3)COP(=O)([O-])[O-])O)O)NC(=NC2=O)N.[Na+].[Na+] | |
407.19 | |
407.19 | |
97% |
Chemical Identifiers
5550-12-9 | |
407.19 | |
PVBRXXAAPNGWGE-LGVAUZIVSA-L | |
21712 | |
C1=NC2=C(N1C3C(C(C(O3)COP(=O)([O-])[O-])O)O)NC(=NC2=O)N.[Na+].[Na+] |
C10H12N5Na2O8P·xH2O | |
MFCD00151238 | |
5'-gmp disodium salt, 5'-guanylic acid disodium salt, 5'-guanylic acid, disodium salt, disodium 5'-gmp, disodium 5'-guanylate, disodium gmp, disodium guanylate, gmp disodium salt, guanosine 5'-monophosphate disodium salt, sodium guanylate | |
disodium;[(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate |
Safety and Handling
EINECSNumber : 226-914-1