missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanine Sulfate Dihydrate 98.0+%, TCI America™
$48.46 - $301.98
Chemical Identifiers
| CAS | 10333-92-3 |
|---|---|
| Molecular Formula | C5H5N5O |
| MDL Number | MFCD00213671 |
| Synonym | 2-Aminohypoxanthine Sulfate |
Chemical Identifiers
| 10333-92-3 | |
| C5H5N5O | |
| MFCD00213671 | |
| 2-Aminohypoxanthine Sulfate |
Specifications
| 10333-92-3 | |
| C5H5N5O | |
| 1 g | |
| 400.33 | |
| Crystalline Powder |
| White | |
| MFCD00213671 | |
| 2-Aminohypoxanthine Sulfate | |
| ≥98.0% (HPLC,N) | |
| Guanine Sulfate Dihydrate |
Safety and Handling
EINECSNumber : (9)-0328
TSCA : Yes