Please login to your online account to display your discounted pricing

Alfa Aesar™ Guanine hydrochloride, 99%

$57.73 - $843.53

Chemical Identifiers

Chemical Name or Material Guanine hydrochloride
CAS 635-39-2
Molecular Formula C5H6ClN5O
Formula Weight 187.59
MDL Number MFCD00213670
Beilstein 4159901
Merck Index 14,4564
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1153218

 
alfa aesar™
A1153218
50g Each for $203.19

AAA1153209

 
alfa aesar
A1153209
10g Each for $57.73

AAA1153230

 
alfa aesar™
A1153230
250g Each for $843.53
Description & Specifications

Specifications

Formula Weight 187.59
Assay Percent Range 99%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Guanine hydrochloride
TSCA TSCA
Assay 99%
Molecular Formula C5H5N5O·HCl
MDL Number MFCD00213670
CAS 635-39-2
Beilstein 4159901
Merck Index 14,4564
Synonym 2-Amino-6-hydroxypurine hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point >300°C
Quantity 50g
Recommended Storage 2°C to 8°C
RTECS Number MF8400000

Applications
Guanine hydrochloride is one of the strongest denaturant. It is mainly used in protein purification and protein folding studies. It is a nucleobase used in biochemical research.

Solubility
Slightly soluble in water

Notes
Store in a cool place. Hygroscopic. Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2°C to 8°C