missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Guanine hydrochloride, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1153218

 View more versions of this product

Catalog No. AAA1153218



Description

Description

Applications
Guanine hydrochloride is one of the strongest denaturant. It is mainly used in protein purification and protein folding studies. It is a nucleobase used in biochemical research.

Solubility
Slightly soluble in water

Notes
Store in a cool place. Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

Guanine hydrochloride
50g
99%
MFCD00213670
14,4564
IBAOFQIOOBQLHE-UHFFFAOYSA-N
2-amino-3,7-dihydropurin-6-one;hydrochloride
69459
99%
>300°C
635-39-2
C5H6ClN5O
4159901
guanine hydrochloride, guanine hcl, 2-amino-1h-purin-6 7h-one hydrochloride, guanine, monohydrochloride, usaf s-1, guanine, hydrochloride, 6h-purin-6-one, 2-amino-1,7-dihydro-, monohydrochloride, 2-amino-9h-purin-6-ol hydrochloride 1:1, 6h-purin-6-one, 2-amino-1,9-dihydro-, hydrochloride 1:1, 2-amino-3,7-dihydropurin-6-one hydrochloride
C1=NC2=C(N1)C(=O)N=C(N2)N.Cl
187.587
187.59

Chemical Identifiers

635-39-2
187.587
IBAOFQIOOBQLHE-UHFFFAOYSA-N
69459
C1=NC2=C(N1)C(=O)N=C(N2)N.Cl
C5H6ClN5O
MFCD00213670
guanine hydrochloride, guanine hcl, 2-amino-1h-purin-6 7h-one hydrochloride, guanine, monohydrochloride, usaf s-1, guanine, hydrochloride, 6h-purin-6-one, 2-amino-1,7-dihydro-, monohydrochloride, 2-amino-9h-purin-6-ol hydrochloride 1:1, 6h-purin-6-one, 2-amino-1,9-dihydro-, hydrochloride 1:1, 2-amino-3,7-dihydropurin-6-one hydrochloride
2-amino-3,7-dihydropurin-6-one;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : MF8400000

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents