missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Guanine, 98%

Manufacturer:  Alfa Aesar™ A1202436

 View more versions of this product

Catalog No. AAA1202436



Description

Description

Applications
Employed as a nucleobase incorporated into biological nucleic acids. Guanine is suitable reagent used to investigate the mechanism of electrochemical oxidation of guanine and adenine using a glassy carbon microelectrode and cyclic and differential pulse voltammetry. It may be used in the preparation of mixed-sequence peptide nucleic acid (PNA) monomers.

Solubility
Insoluble in water. Soluble in ammonia-water, KOH solutions, dilute acids, and 1 N NaOH (0.1M).

Notes
Store away from oxidizing agents. Keep the container tightly closed in a well ventilated place.
Specifications

Specifications

Guanine
98%
MFCD00071533
14,4564
UYTPUPDQBNUYGX-UHFFFAOYSA-N
2-amino-3,7-dihydropurin-6-one
764
151.13
73-40-5
C5H5N5O
9680
2-Amino-6-hydroxypurine
C1=NC2=C(N1)C(=O)N=C(N2)N
151.129
CHEBI:16235

Chemical Identifiers

73-40-5
151.129
UYTPUPDQBNUYGX-UHFFFAOYSA-N
764
2-amino-3,7-dihydropurin-6-one
C5H5N5O
MFCD00071533
2-Amino-6-hydroxypurine
CHEBI:16235
C1=NC2=C(N1)C(=O)N=C(N2)N
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : MF8260000

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents